Learn More
Medchemexpress LLC HY-128892 5mg , EN6 CAS:1808714-73-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1288925MG
Encompass_Preferred
HY-128892 5mg EN6 CAS: 1808714-73-9 EN6 is a small-molecule in vivo activator of autophagy that covalently targets cysteine 277 in the ATP6V1A subunit of the lysosomal the vacuolar H+ ATPase (v-ATPase). EN6-mediated ATP6V1A modification decouples the v-ATPase from the Rags, leading to inhibition of mTORC1 signaling, increased lysosomal acidification and activation of autophagy. EN6 clears TDP-43 aggregates, a causative agent in frontotemporal dementia, in a lysosome-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0674