Learn More
TARGETMOL CHEMICALS INC Enalapril 100MG
SDP
Catalog No. 502877548
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Enalapril is a dicarbocyl-containing peptide and angiotensin-converting enzyme (ACE) inhibitor with antihypertensive activity. Purity 99.47%
Catalog No. 50-287-7548 Supplier TARGETMOL CHEMICALS INC Supplier No. T1605100MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Enalapril is a dicarbocyl-containing peptide and angiotensin-converting enzyme (ACE) inhibitor with antihypertensive activity. Purity 99.47%