Learn More
Selleck Chemical LLC Enalapril E4900-1G
SDP
Catalog No. 5000041094
Encompass_Preferred
Enalapril(Eletriptan Amprace Vaseretic) is an inhibitor of angiotensin-converting enzyme (ACE) primarily used to treat hypertension heart failure and asymptomatic left ventricular dysfunction It also attenuates frailty in aging mice
Catalog No. 50-000-41094 Supplier Selleck Chemical LLC Supplier No. E49001G
Enalapril(Eletriptan Amprace Vaseretic) is an inhibitor of angiotensin-converting enzyme (ACE) primarily used to treat hypertension heart failure and asymptomatic left ventricular dysfunction It also attenuates frailty in aging mice