Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enalapril | 75847-73-3 | MFCD00865774 | 99.96% | 50 MG
SDP

Catalog No. 5000406678
Encompass_Preferred

Enalapril (MK-421) is an angiotensin-converting enzyme (ACE) inhibitor commonly used in hypertensive diseases research. It is a solid, white to off-white compound with a high purity of 99.96%.

  • Angiotensin-converting enzyme (ACE) inhibitor
  • Suitable for hypertensive diseases research
  • High purity of 99.96%
  • Solid, white to off-white appearance
  • Soluble in DMSO and water
  • Melting point of 142-147 °C
  • Store under inert gas (nitrogen or argon) at 2-8 °C

Catalog No. 50-004-06678 Supplier Medchemexpress LLC Supplier No. HYB033150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enalapril (MK-421) is an angiotensin-converting enzyme (ACE) inhibitor commonly used in hypertensive diseases research. It is a solid, white to off-white compound with a high purity of 99.96%.

  • Angiotensin-converting enzyme (ACE) inhibitor
  • Suitable for hypertensive diseases research
  • High purity of 99.96%
  • Solid, white to off-white appearance
  • Soluble in DMSO and water
  • Melting point of 142-147 °C
  • Store under inert gas (nitrogen or argon) at 2-8 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.