Learn More
Medchemexpress LLC Enalapril-d5 (maleate) | 349554-02-5 | 99.0% | 1 MG
SDP
Catalog No. 5000406923
Encompass_Preferred
Enalapril-d5 (maleate) is deuterium labeled Enalapril, an angiotensin converting enzyme (ACE) inhibitor. Intended for research use only, this compound serves as a valuable tool for quantitative analysis.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06923 Supplier Medchemexpress LLC Supplier No. HYB0331AS1MG
Enalapril-d5 (maleate) is deuterium labeled Enalapril, an angiotensin converting enzyme (ACE) inhibitor. Intended for research use only, this compound serves as a valuable tool for quantitative analysis.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS