Learn More
Medchemexpress LLC Enalapril-d5 (maleate) | 349554-02-5 | M001579 | 99.0% | 5 MG
SDP
Catalog No. 5000406377
Encompass_Preferred
Enalapril-d5 maleate is a deuterium-labeled form of Enalapril, a potent and long-acting angiotensin-converting enzyme (ACE) inhibitor.
  • Used for treating hypertension, congestive heart failure, and diabetic nephropathy.
  • Inhibits the conversion of angiotensin I to angiotensin II.
  • Results in decreased vasoconstriction and reduced aldosterone secretion.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06377 Supplier Medchemexpress LLC Supplier No. HYB0331AS5MG
Enalapril-d5 maleate is a deuterium-labeled form of Enalapril, a potent and long-acting angiotensin-converting enzyme (ACE) inhibitor.
  • Used for treating hypertension, congestive heart failure, and diabetic nephropathy.
  • Inhibits the conversion of angiotensin I to angiotensin II.
  • Results in decreased vasoconstriction and reduced aldosterone secretion.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.