Learn More
Apexbio Technology LLC Enalapril Maleate 76095-16-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062080
Encompass_Preferred
Enalapril Maleate (CAS 76095-16-4) is the maleate salt of enalapril a prodrug metabolized in vivo into its active metabolite enalaprilat It exerts its pharmacological activity through inhibition of angiotensin-converting enzyme (ACE) thereby preventing conversion of angiotensin I into angiotensin II This action reduces aldosterone secretion and attenuates vasoconstriction Enalapril Maleate is widely utilized in basic and clinical cardiovascular research particularly for examining the renin-angiotensin system (RAS) and associated cardiovascular pathophysiology
Catalog No. 50-000-62080 Supplier Apexbio Technology LLC Supplier No. B22055.1
Enalapril Maleate (CAS 76095-16-4) is the maleate salt of enalapril a prodrug metabolized in vivo into its active metabolite enalaprilat It exerts its pharmacological activity through inhibition of angiotensin-converting enzyme (ACE) thereby preventing conversion of angiotensin I into angiotensin II This action reduces aldosterone secretion and attenuates vasoconstriction Enalapril Maleate is widely utilized in basic and clinical cardiovascular research particularly for examining the renin-angiotensin system (RAS) and associated cardiovascular pathophysiology