Learn More
Medchemexpress LLC Enalapril maleate | 76095-16-4 | 100.0% | 1 ML
SDP
Catalog No. 5000406709
Encompass_Preferred
Enalapril maleate is an active metabolite of enalapril and a potent angiotensin-converting enzyme (ACE) inhibitor. As a prodrug, it is rapidly metabolized to enalaprilat, which helps regulate blood pressure by targeting the renin-angiotensin-aldosterone system (RAAS). It is used in the treatment of essential or renovascular hypertension and symptomatic congestive heart failure.
  • Active metabolite of enalapril
  • Angiotensin-converting enzyme (ACE) inhibitor
  • Prodrug that converts to enalaprilat
  • Potent and competitive ACE inhibitor
  • Used to treat hypertension
  • Used to treat congestive heart failure
Catalog No. 50-004-06709 Supplier Medchemexpress LLC Supplier No. HYB0331A10MM/1ML
Enalapril maleate is an active metabolite of enalapril and a potent angiotensin-converting enzyme (ACE) inhibitor. As a prodrug, it is rapidly metabolized to enalaprilat, which helps regulate blood pressure by targeting the renin-angiotensin-aldosterone system (RAAS). It is used in the treatment of essential or renovascular hypertension and symptomatic congestive heart failure.
  • Active metabolite of enalapril
  • Angiotensin-converting enzyme (ACE) inhibitor
  • Prodrug that converts to enalaprilat
  • Potent and competitive ACE inhibitor
  • Used to treat hypertension
  • Used to treat congestive heart failure