Learn More
Medchemexpress LLC Enalapril (maleate) | 76095-16-4 | 99.95% | 250 MG
SDP
Catalog No. 5000406163
Encompass_Preferred
Enalapril maleate, also known as MK-421 maleate, is the active metabolite of enalapril and functions as an angiotensin-converting enzyme (ACE) inhibitor. Enalapril is a prodrug that is rapidly metabolized in the liver to enalaprilat following oral administration. It is a potent, competitive inhibitor of ACE, which is responsible for converting angiotensin I to angiotensin II, a key regulator of blood pressure. Enalapril may be used to treat essential or renovascular hypertension and symptomatic congestive heart failure. This product is for research use only.
  • Harmful if swallowed.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Avoid breathing dust, fume, gas, mist, vapors, or spray.
  • Keep container tightly sealed in a cool, well-ventilated area, away from moisture and direct sunlight.
  • Wear protective gloves, clothing, eye protection, and face protection.
Catalog No. 50-004-06163 Supplier Medchemexpress LLC Supplier No. 250MGHYB0331AR
Enalapril maleate, also known as MK-421 maleate, is the active metabolite of enalapril and functions as an angiotensin-converting enzyme (ACE) inhibitor. Enalapril is a prodrug that is rapidly metabolized in the liver to enalaprilat following oral administration. It is a potent, competitive inhibitor of ACE, which is responsible for converting angiotensin I to angiotensin II, a key regulator of blood pressure. Enalapril may be used to treat essential or renovascular hypertension and symptomatic congestive heart failure. This product is for research use only.
  • Harmful if swallowed.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Avoid breathing dust, fume, gas, mist, vapors, or spray.
  • Keep container tightly sealed in a cool, well-ventilated area, away from moisture and direct sunlight.
  • Wear protective gloves, clothing, eye protection, and face protection.