Learn More
Medchemexpress LLC Enalapril (maleate) | 76095-16-4 | 99.99% | 5 G
SDP
Catalog No. 5000406680
Encompass_Preferred
Enalapril (MK-421) maleate is an angiotensin-converting enzyme (ACE) inhibitor and the active metabolite of enalapril. This prodrug is rapidly metabolized in the liver to enalaprilat after oral administration. It is a potent, competitive inhibitor of ACE, an enzyme crucial for converting angiotensin I to angiotensin II, which regulates blood pressure and is a key component of the renin-angiotensin-aldosterone system (RAAS).
  • Treats essential or renovascular hypertension
  • Treats symptomatic congestive heart failure
  • Functions as a potent, competitive ACE inhibitor
  • Metabolized to enalaprilat in the liver
  • For research use only
Catalog No. 50-004-06680 Supplier Medchemexpress LLC Supplier No. HYB0331A5G
Enalapril (MK-421) maleate is an angiotensin-converting enzyme (ACE) inhibitor and the active metabolite of enalapril. This prodrug is rapidly metabolized in the liver to enalaprilat after oral administration. It is a potent, competitive inhibitor of ACE, an enzyme crucial for converting angiotensin I to angiotensin II, which regulates blood pressure and is a key component of the renin-angiotensin-aldosterone system (RAAS).
  • Treats essential or renovascular hypertension
  • Treats symptomatic congestive heart failure
  • Functions as a potent, competitive ACE inhibitor
  • Metabolized to enalaprilat in the liver
  • For research use only