Learn More
Selleck Chemical LLC Enalapril Maleate 100mg 76095-16-4 MK-421 Maleate
SDP
Catalog No. 505556
Encompass_Preferred
Enalapril (MK-421) Maleate is an angiotensin-converting enzyme (ACE) inhibitor, used in the treatment of hypertension, diabetic nephropathy, and chronic heart failure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-555-6 Supplier Selleck Chemical LLC Supplier No. S1941100MG
Enalapril (MK-421) Maleate is an angiotensin-converting enzyme (ACE) inhibitor, used in the treatment of hypertension, diabetic nephropathy, and chronic heart failure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.