Learn More
Medchemexpress LLC Enalapril maleate (Standard) | 76095-16-4 | 100.0% | 25 MG
SDP
Catalog No. 5000450530
Encompass_Preferred
Enalapril maleate (Standard) is an analytical standard and an angiotensin-converting enzyme (ACE) inhibitor. This product is intended for research and analytical applications, serving as a reference standard for assays.
  • Reference standard for assay
  • Used in qualitative, quantitative, and methodological research
  • Applicable for HPLC, GC, and MS applications
Catalog No. 50-004-50530 Supplier Medchemexpress LLC Supplier No. 25MGHYB0331AR
Enalapril maleate (Standard) is an analytical standard and an angiotensin-converting enzyme (ACE) inhibitor. This product is intended for research and analytical applications, serving as a reference standard for assays.
  • Reference standard for assay
  • Used in qualitative, quantitative, and methodological research
  • Applicable for HPLC, GC, and MS applications