Learn More
Medchemexpress LLC Enalapril maleate (standard) | 76095-16-4 | 99.95% | 100 MG
SDP
Catalog No. 5000406379
Encompass_Preferred
Enalapril maleate (Standard) is an analytical standard of Enalapril maleate intended for research and analytical applications. This product, also known as MK-421 maleate, is the active metabolite of enalapril and acts as an angiotensin-converting enzyme (ACE) inhibitor. It is a reference standard for assaying and is commonly utilized in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Serves as an analytical standard
  • Suitable for research and analytical applications
  • Functions as an angiotensin-converting enzyme (ACE) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments
  • Compatible with HPLC, GC, and MS techniques
Catalog No. 50-004-06379 Supplier Medchemexpress LLC Supplier No. HYB0331AR100MG
Enalapril maleate (Standard) is an analytical standard of Enalapril maleate intended for research and analytical applications. This product, also known as MK-421 maleate, is the active metabolite of enalapril and acts as an angiotensin-converting enzyme (ACE) inhibitor. It is a reference standard for assaying and is commonly utilized in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Serves as an analytical standard
  • Suitable for research and analytical applications
  • Functions as an angiotensin-converting enzyme (ACE) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments
  • Compatible with HPLC, GC, and MS techniques