Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enalapril maleate (Standard) | 76095-16-4 | 99.95% | 50 MG
SDP

Catalog No. 5000405859
Encompass_Preferred

Enalapril maleate (Standard) is an analytical standard and an angiotensin-converting enzyme (ACE) inhibitor. This product is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological experiments such as HPLC, GC, and MS.

  • Analytical standard for research and analytical applications
  • Angiotensin-converting enzyme (ACE) inhibitor
  • Suitable for HPLC, GC, and MS research experiments
  • Molecular weight: 492.52
  • Formula: C₂₄H₃₂N₂O₉
  • Appearance: solid
  • Color: white to off-white

Catalog No. 50-004-05859 Supplier Medchemexpress LLC Supplier No. HYB0331AR50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enalapril maleate (Standard) is an analytical standard and an angiotensin-converting enzyme (ACE) inhibitor. This product is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological experiments such as HPLC, GC, and MS.

  • Analytical standard for research and analytical applications
  • Angiotensin-converting enzyme (ACE) inhibitor
  • Suitable for HPLC, GC, and MS research experiments
  • Molecular weight: 492.52
  • Formula: C₂₄H₃₂N₂O₉
  • Appearance: solid
  • Color: white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.