Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enalapril (MK-421) | 75847-73-3 | 99.96% | 1 ML
SDP

Catalog No. 5000406554
Encompass_Preferred

Enalapril (MK-421) is an angiotensin-converting enzyme (ACE) inhibitor that can be used for hypertensive diseases research.

  • CAS number is 75847-73-3.
  • Purity is 99.96%.
  • Molecular weight is 376.45.
  • Chemical formula is C20H28N2O5.
  • Appears as a white to off-white solid.
  • Soluble in DMSO and water.
  • Used for hypertensive diseases research.
  • Exhibits antiarrhythmic effect in canine hearts.
  • Inhibits apoptosis in human umbilical vein endothelial cells with Alzheimer's disease serum.
  • Reduces infarct volume in male NMRI mice at 0.03 mg/kg.

Catalog No. 50-004-06554 Supplier Medchemexpress LLC Supplier No. HYB033110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enalapril (MK-421) is an angiotensin-converting enzyme (ACE) inhibitor that can be used for hypertensive diseases research.

  • CAS number is 75847-73-3.
  • Purity is 99.96%.
  • Molecular weight is 376.45.
  • Chemical formula is C20H28N2O5.
  • Appears as a white to off-white solid.
  • Soluble in DMSO and water.
  • Used for hypertensive diseases research.
  • Exhibits antiarrhythmic effect in canine hearts.
  • Inhibits apoptosis in human umbilical vein endothelial cells with Alzheimer's disease serum.
  • Reduces infarct volume in male NMRI mice at 0.03 mg/kg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.