Learn More
Abcam Enalaprilat, Angiotensin II converting enzyme inhibitor, 50MG
Catalog No. 502706154
Encompass_Preferred
MW 384.4 Da, Purity >99%. Angiotensin II converting enzyme inhibitor. Inhibits angiotensin II-induced cardiac fibroblast proliferation. Active metabolite of Enalapril (ab141929). Antihypertensive agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6154 Supplier Abcam Supplier No. AB14286650MG
Add to Cart
Add to Cart
MW 384.4 Da, Purity >99%. Angiotensin II converting enzyme inhibitor. Inhibits angiotensin II-induced cardiac fibroblast proliferation. Active metabolite of Enalapril (ab141929). Antihypertensive agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement