Learn More
Medchemexpress LLC Enalaprilat-d5 | 349554-00-3 | 99.5% | 1 MG
SDP
Catalog No. 5000405494
Encompass_Preferred
Enalaprilat-d5 is a deuterium-labeled form of Enalaprilat (MK-422), functioning as an angiotensin-converting enzyme (ACE) inhibitor. It can be utilized as a tracer and as an internal standard in various quantitative analyses.
  • Deuterium-labeled ACE inhibitor
  • Can be used as a tracer
  • Internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Targets angiotensin-converting enzyme (ACE)
  • Involved in metabolic enzyme/protease pathway
Catalog No. 50-004-05494 Supplier Medchemexpress LLC Supplier No. HYB0231AS1MG
Enalaprilat-d5 is a deuterium-labeled form of Enalaprilat (MK-422), functioning as an angiotensin-converting enzyme (ACE) inhibitor. It can be utilized as a tracer and as an internal standard in various quantitative analyses.
  • Deuterium-labeled ACE inhibitor
  • Can be used as a tracer
  • Internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Targets angiotensin-converting enzyme (ACE)
  • Involved in metabolic enzyme/protease pathway