Learn More
Medchemexpress LLC Enalaprilat-d5 | 349554-00-3 | 99.5% | 5 MG
SDP
Catalog No. 5000406262
Encompass_Preferred
Enalaprilat-d5 is the deuterium labeled Enalaprilat (MK-422), an angiotensin-converting enzyme (ACE) inhibitor. This compound is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06262 Supplier Medchemexpress LLC Supplier No. HYB0231AS5MG
Add to Cart
Add to Cart
Enalaprilat-d5 is the deuterium labeled Enalaprilat (MK-422), an angiotensin-converting enzyme (ACE) inhibitor. This compound is for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS