Learn More
Cayman Chemical EnalaprIlathydrate 100mg
SDP
Catalog No. 502965454
Encompass_Preferred
The active metabolite of the ACE inhibitor, enalapril; inhibits ACE activity with an IC50 value of 5.8 nM in vitro
Catalog No. 50-296-5454 Supplier Cayman Chemical Supplier No. 17603100 MG
The active metabolite of the ACE inhibitor, enalapril; inhibits ACE activity with an IC50 value of 5.8 nM in vitro