Learn More
Cayman Chemical Enalaprilat (hydrate), Purity: ≥98%, CAS: 84680-54-6, 50 mg
SDP
Catalog No. 502965691
Encompass_Preferred
The active metabolite of the ACE inhibitor, enalapril; inhibits ACE activity with an IC50 value of 5.8 nM in vitro
Catalog No. 50-296-5691 Supplier Cayman Chemical Supplier No. 1760350 MG
The active metabolite of the ACE inhibitor, enalapril; inhibits ACE activity with an IC50 value of 5.8 nM in vitro