Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Enantioprobe (R R)-7

Catalog No. 91341350MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-341-350MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-341-350MG Supplier Sigma Aldrich Supplier No. 91341350MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Supporting reagents:Fluorophore-conjugated azide tags for SDS-PAGE in-gel profiling: 910147, 760757, 760765, 777315, 777323Azide-biotin tags for enrichment: 901765, 914134, 914460, 762024, 900912, 900891, QBD10825Azide capture solid supports: 900957, 742627Streptavidin for biotin enrichment: S6940, E5529, E2513, 69203, 16-126, S1638 Mass spectrometry and MultiplexingExpedited mapping of the ligandable proteome using fully functionalized enantiomeric probe pairsA chiral trick to map protein ligandability

Specifications

Molecular Formula C7H4N2O5S
Linear Formula C7H4N2O5S
Quantity 1 g
Synonym 2-Nitrobenzo[d]isothiazolone 1,1-dioxide
Molecular Weight (g/mol) 228.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.