Learn More
Cayman Chemical Enarodustat 10MG
SDP
Catalog No. 502949795
Encompass_Preferred
A pan HIF-PH inhibitor (Kis = 16, 61, and 101 nM for HIF-PH1, -2, and -3, respectively); increases EPO levels in Hep3B cells (EC50 = 4.7 µM); increases plasma EPO levels in control rats, as well as plasma EPO and hemoglobin levels in 5/6-nephrectomized rats at 3 mg/kg; decreases renal fatty acid and amino acid metabolism and increases renal glucose metabolism in streptozotocin-induced rat and alloxan-induced mouse models of diabetic nephropathy
Catalog No. 50-294-9795 Supplier Cayman Chemical Supplier No. 3378510 MG
A pan HIF-PH inhibitor (Kis = 16, 61, and 101 nM for HIF-PH1, -2, and -3, respectively); increases EPO levels in Hep3B cells (EC50 = 4.7 µM); increases plasma EPO levels in control rats, as well as plasma EPO and hemoglobin levels in 5/6-nephrectomized rats at 3 mg/kg; decreases renal fatty acid and amino acid metabolism and increases renal glucose metabolism in streptozotocin-induced rat and alloxan-induced mouse models of diabetic nephropathy