Learn More
Medchemexpress LLC HY-18690 50mg , Enasidenib CAS:1446502-11-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1869050MG
Encompass_Preferred
HY-18690 50mg Enasidenib CAS: 1446502-11-9 Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1023