Learn More
Medchemexpress LLC Enasidenib mesylate | 1650550-25-6 | 99.7% | 569.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY18690A
Encompass_Preferred
Enasidenib mesylate is a first-in-class, oral, potent, reversible, and selective inhibitor of the IDH2 mutant enzymes. It reverses the effects of mutant IDH2 on DNA methylation in mutant stem/progenitor cells, inducing differentiation and impairing self-renewal of IDH2-mutant leukemia cells. In vivo studies demonstrate that treatment with enasidenib significantly improves survival in an IDH2-mutant acute myeloid leukemia (AML) primary xenograft mouse model by remodeling the epigenetic state and reducing 2-hydroxyglutarate (2-HG) levels, consistent with on-target inhibition.
  • Novel approach to treatment
  • Convenient oral administration
  • Potent, reversible, and selective inhibitor
  • Induces differentiation of leukemic cells
  • Improves survival in AML models
  • Effectively reduces 2-hydroxyglutarate
  • Remodels epigenetic state of IDH2-mutant cells
Catalog No. 50-003-93392