Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics eNat™ 6U073S01
SDP

Catalog No. 23600987 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
23-600-987 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-600-987 Supplier Copan Diagnostics Supplier No. 6U073S01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Regular flocked swab and eNat™ medium

  • 80 mm breakpoint
  • Packaged with 2 mL eNat™ Guanidine Medium in skirted tube with plastic cap
  • Individually packaged
  • Sterile

Specifications

Includes 2 mL eNat™ guanidine medium in skirted tube with plastic cap
Sterility Sterile
Packaging Individually packaged
For Use With (Application) Specimen collection and transport for molecular assays
Breakpoint 80 mm
Product Type Regular flocked swab, eNat™ medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.