Learn More
Medchemexpress LLC Enavogliflozin | 1415472-28-4 | 98.4% | 446.92 g·mol⁻¹ | C24H27ClO6 | 5 MG
SDP
Catalog No. 5000264021
Encompass_Preferred
Enavogliflozin is an orally active, selective sodium-glucose cotransporter-2 (SGLT-2) inhibitor developed for treatment of type 2 diabetes and related metabolic disorders. The compound is provided to researchers as a solid powder and as concentrated solution formats for in vitro and translational studies, and it has been characterized in peer-reviewed studies and regulatory entries.
Catalog No. 50-002-64021 Supplier Medchemexpress LLC Supplier No. HY1091445MG
Enavogliflozin is an orally active, selective sodium-glucose cotransporter-2 (SGLT-2) inhibitor developed for treatment of type 2 diabetes and related metabolic disorders. The compound is provided to researchers as a solid powder and as concentrated solution formats for in vitro and translational studies, and it has been characterized in peer-reviewed studies and regulatory entries.