Learn More
Medchemexpress LLC Enbezotinib | 2359649-81-1 | 99.5% | C21H21FN6O3 | 100 MG
SDP
Catalog No. 5000356087
Encompass_Preferred
Enbezotinib, also known as TPX-0046, is an inhibitor of RET that can inhibit RET autophosphorylation and is utilized for cancer research.
  • Inhibits p-RET (y905) in RET-driven cells at concentrations of 0.3-300 nM in vitro
  • Decreases tumor size in mice when administered at 2-5 mg/kg, twice daily for 27 days in vivo
  • Molecular weight: 424.43
Catalog No. 50-003-56087 Supplier Medchemexpress LLC Supplier No. HY145565100MG
Enbezotinib, also known as TPX-0046, is an inhibitor of RET that can inhibit RET autophosphorylation and is utilized for cancer research.
  • Inhibits p-RET (y905) in RET-driven cells at concentrations of 0.3-300 nM in vitro
  • Decreases tumor size in mice when administered at 2-5 mg/kg, twice daily for 27 days in vivo
  • Molecular weight: 424.43