Learn More
Medchemexpress LLC Enbezotinib | 2359649-81-1 | 99.5% | C21H21FN6O3 | 50 MG
SDP
Catalog No. 5000355273
Encompass_Preferred
Enbezotinib, also known as TPX-0046, is a potent inhibitor of RET (rearranged during transfection) kinase. It functions by inhibiting RET autophosphorylation, a key process involved in various cellular activities, including cell growth, differentiation, and survival. This compound is primarily utilized in cancer research due to its ability to target and suppress RET activity, which is often aberrantly activated in several types of cancer.
  • Inhibits RET autophosphorylation
  • Used for cancer research
  • Potent RET kinase inhibitor
Catalog No. 50-003-55273 Supplier Medchemexpress LLC Supplier No. HY14556550MG
Enbezotinib, also known as TPX-0046, is a potent inhibitor of RET (rearranged during transfection) kinase. It functions by inhibiting RET autophosphorylation, a key process involved in various cellular activities, including cell growth, differentiation, and survival. This compound is primarily utilized in cancer research due to its ability to target and suppress RET activity, which is often aberrantly activated in several types of cancer.
  • Inhibits RET autophosphorylation
  • Used for cancer research
  • Potent RET kinase inhibitor