Learn More
Medchemexpress LLC Enbezotinib (TPX-0046) | 2359649-81-1 | 99.5% | 424.43 g/mol | C21H21FN6O3 | 5 MG
SDP
Catalog No. 5000221702
Encompass_Preferred
Enbezotinib is a small-molecule RET kinase inhibitor for preclinical cancer research. It inhibits RET autophosphorylation, exhibits potent in vitro activity against RET-driven cells, and has demonstrated tumor size reduction in animal models. Supplied as a high-purity solid with solution formats available for laboratory use.
  • Inhibits RET autophosphorylation.
  • Potent in vitro activity against RET-driven cells.
  • Demonstrated tumor growth reduction in animal models.
  • High-purity solid and solution formats available.
  • Intended for research use only.
Catalog No. 50-002-21702 Supplier Medchemexpress LLC Supplier No. HY1455655MG
Enbezotinib is a small-molecule RET kinase inhibitor for preclinical cancer research. It inhibits RET autophosphorylation, exhibits potent in vitro activity against RET-driven cells, and has demonstrated tumor size reduction in animal models. Supplied as a high-purity solid with solution formats available for laboratory use.
  • Inhibits RET autophosphorylation.
  • Potent in vitro activity against RET-driven cells.
  • Demonstrated tumor growth reduction in animal models.
  • High-purity solid and solution formats available.
  • Intended for research use only.