Learn More
Medchemexpress LLC Enbezotinib (TPX-0046) | 2359649-81-1 | 99.5% | C21H21FN6O3 | 10MG
SDP
Catalog No. 5000259790
Encompass_Preferred
Enbezotinib is a small-molecule RET kinase inhibitor used in preclinical cancer research. It inhibits RET autophosphorylation and is applied in studies of RET-driven tumors and signaling pathways. Supplied as a solid, it is intended for biochemical and cellular assay applications requiring a defined chemical standard.
  • Inhibits RET autophosphorylation for RET-targeted research.
  • High purity suitable for biochemical and cellular assays.
  • Solid form convenient for storage and handling.
  • Molecular weight 424.4 g/mol and defined formula for analytical use.
  • Useful in preclinical studies of RET-driven cancers.
Catalog No. 50-002-59790 Supplier Medchemexpress LLC Supplier No. HY14556510MG
Enbezotinib is a small-molecule RET kinase inhibitor used in preclinical cancer research. It inhibits RET autophosphorylation and is applied in studies of RET-driven tumors and signaling pathways. Supplied as a solid, it is intended for biochemical and cellular assay applications requiring a defined chemical standard.
  • Inhibits RET autophosphorylation for RET-targeted research.
  • High purity suitable for biochemical and cellular assays.
  • Solid form convenient for storage and handling.
  • Molecular weight 424.4 g/mol and defined formula for analytical use.
  • Useful in preclinical studies of RET-driven cancers.