Learn More
Hach Company Encap Diff. pH Sens-LCP-Preamp
Catalog No. 5000207708 Shop All Hach Company Products
Encompass
Encapsulated Differential pH Sensor-LCP Body-5-Wire with Internal Preamplifier-Glass Electrode-90 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 27 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode 90 ft Cable
Catalog No. 50-002-07708 Supplier Hach Company Supplier No. 6028P090
Encapsulated Differential pH Sensor-LCP Body-5-Wire with Internal Preamplifier-Glass Electrode-90 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 27 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode 90 ft Cable