Learn More
Hach Company Encap Diff. pH Sens-LCP-Preamp
Catalog No. 5000207814 Shop All Hach Company Products
Encompass
Encapsulated LCP (Liquid Crystal Polymer) Differential pH Sensor-5-Wire with Internal Preamplifier-Glass Electrode-33ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 10m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07814 Supplier Hach Company Supplier No. 6028P033
Encapsulated LCP (Liquid Crystal Polymer) Differential pH Sensor-5-Wire with Internal Preamplifier-Glass Electrode-33ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 10m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode