Learn More
Hach Company Encap Diff. pH Sens-LCP-Preamp
Catalog No. 5000207870 Shop All Hach Company Products
Encompass
Encapsulated LCP Differential pH Sensor-Internal Preamplifier-Glass Electrode-5-Wire-15 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 4 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07870 Supplier Hach Company Supplier No. 6028P015
Encapsulated LCP Differential pH Sensor-Internal Preamplifier-Glass Electrode-5-Wire-15 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 4 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode