Learn More
Hach Company Encap PPS Diffl pH Sens-5-Wire
Catalog No. 5000207579 Shop All Hach Company Products
Encompass
Encapsulated PPS Differential pH Sensor-5-Wire with Internal Preamplifier and Glass Electrode-10 ft cablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07579 Supplier Hach Company Supplier No. 6022P0
Encapsulated PPS Differential pH Sensor-5-Wire with Internal Preamplifier and Glass Electrode-10 ft cablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode