Learn More
Hach Company Encap PPS pH Sens-LCP-5-Wire
Catalog No. 5000208095 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Amphenol Termination-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Cicular MIL Connector - 7 Pin
Catalog No. 50-002-08095 Supplier Hach Company Supplier No. 6028P01000A000N
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Amphenol Termination-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Cicular MIL Connector - 7 Pin