Learn More
Hach Company Encap PPS pH Sens-LCP-5-Wire
Catalog No. 5000207840 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Salt Bridge-Perflouroelastomer O-RingsLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode FFKM/FFPM O-Rings
Catalog No. 50-002-07840 Supplier Hach Company Supplier No. 6028P010000200N
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Salt Bridge-Perflouroelastomer O-RingsLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode FFKM/FFPM O-Rings