Learn More
Sigma Aldrich EncapGel-Cx Kit, chemically crosslinkable sIPN hydrogel kit
Catalog No. 7996291KT Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kt
Catalog No. Quantity
79-962-91KT 1 kt
1 options
Catalog No. 79-962-91KT Supplier Sigma Aldrich Supplier No. 7996291KT
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
EncapGel(TM)-Cx Kit is a ready-to-use, chemically crosslinked hydrogel kit for the encapsulation and delivery of drugs, biomolecules, and cells. This hydrogel fuses the durability of synthetic polymers and the biorecognition of natural proteins to create a biocompatible delivery platform. The biodegradable matrix allows for temporally and spacially controlled delivery. Applications include:Localized delivery of small molecules, proteins, nucleic acids, and cellular-based therapeutics Tissue engineeringRegenerative medicineENCAPGEL is a trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 kt