Learn More
ENCAPSULA NANAOSCIENCE LLC Encapsome 10mL
Catalog No. NC1676287
Encompass
Control liposomes (Encapsome) are recognized and phagocytosed by the same mechanism as Clodrosome. Since the control liposomes do not contain clodronate, the phagocytic cells are not killed. However, phagocytes do respond to the ingestion of control liposomes by cytokine secretion, temporary suspension of phagocytic activity.
Catalog No. NC1676287 Supplier ENCAPSULA NANAOSCIENCE LLC Supplier No. CLD891010ML
Add to Cart
Add to Cart
Control liposomes (Encapsome) are recognized and phagocytosed by the same mechanism as Clodrosome. Since the control liposomes do not contain clodronate, the phagocytic cells are not killed. However, phagocytes do respond to the ingestion of control liposomes by cytokine secretion, temporary suspension of phagocytic activity.