Learn More
Perkin Elmer US LLC Encapsulated Kit -Calibration Materials Kit
Catalog No. 502112122
Encompass_Preferred
The reference materials in the Encapsulated Kit are 99.999% pure. DSC instruments require elements of this purity to ensure accurate calibrations.
Catalog No. 50-211-2122 Supplier Perkin Elmer US LLC Supplier No. 02190045
The reference materials in the Encapsulated Kit are 99.999% pure. DSC instruments require elements of this purity to ensure accurate calibrations.