Learn More
V&P Scientific Inc STIR ELEMENT, DISC, SmCo Magnet, 29MGO, PTFE Encapsulated, 5.0mm Diameter, 1.7mm Height, 12/pack
Catalog No. NC1635825
Encompass
STIR ELEMENT, DISC, SmCo Magnet, 29MGO, PTFE Encapsulated, 5.0mm Diameter, 1.7mm Height, 260°C Max Operating Temp for Coating
Catalog No. NC1635825 Supplier V&P Scientific Inc Supplier No. VP 7795
STIR ELEMENT, DISC, SmCo Magnet, 29MGO, PTFE Encapsulated, 5.0mm Diameter, 1.7mm Height, 260°C Max Operating Temp for Coating