Learn More
Genscript Corporation Enceed T cell Activation hum
Supplier:  Genscript Corporation L00935
Encompass
The Enceed T cell Activation human (GMP) is used for in vitro activation and expansion of enriched T cells or peripheral blood mononuclear cells (PBMCs) Due to the nanoscale structure of Enceed T cell Activation human (GMP) the excess reagent can be removed by centrifugation or conventional supernatant replacement
Catalog No. 50-000-57102