Learn More
Genscript Corporation Enceed T cell Activation hum
Catalog No. 5000057426
Encompass
The Enceed T cell Activation reagent induces T cell activation and expansion in enriched T cells or PBMCs The Enceed T cell Activation reagent is composed of biodegradable matrix-coated nanoparticles conjugated with mouse anti-human CD3 and mouse anti-human CD28 antibodies The small sized particulate structure of the activation reagent enables its easy removal by media washing or centrifugation
Catalog No. 50-000-57426 Supplier Genscript Corporation Supplier No. L008991
The Enceed T cell Activation reagent induces T cell activation and expansion in enriched T cells or PBMCs The Enceed T cell Activation reagent is composed of biodegradable matrix-coated nanoparticles conjugated with mouse anti-human CD3 and mouse anti-human CD28 antibodies The small sized particulate structure of the activation reagent enables its easy removal by media washing or centrifugation