Learn More
Selleck Chemical LLC Encenicline (EVP-6124) hydrochloride S2957-5mg
SDP
Catalog No. 503111759
Encompass_Preferred
Encenicline (EVP-6124) hydrochloride is a selective 7 nicotinic acetylcholine receptor partial agonist improves memory performance by potentiating the acetylcholine response of 7 nicotinic acetylcholine receptors
Catalog No. 50-311-1759 Supplier Selleck Chemical LLC Supplier No. S29575MG
Encenicline (EVP-6124) hydrochloride is a selective 7 nicotinic acetylcholine receptor partial agonist improves memory performance by potentiating the acetylcholine response of 7 nicotinic acetylcholine receptors