Learn More
Medchemexpress LLC Encenicline hydrochloride | 550999-74-1 | C16H18Cl2N2OS | 100 MG
SDP
Catalog No. 5000376226
Encompass_Preferred
Encenicline hydrochloride is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs). This compound is intended for research use only and is not sold to patients. It demonstrates selectivity for α7 nAChRs and has shown promising results in restoring memory function in preclinical studies.
  • Novel partial agonist of α7 neuronal nicotinic acetylcholine receptors
  • Purity of 98.01%
  • White to off-white solid appearance
  • Exhibits good brain penetration
  • Restores memory function in scopolamine-treated rats
  • Moderately binds to rat plasma proteins
Catalog No. 50-003-76226 Supplier Medchemexpress LLC Supplier No. HY15430A100MG
Encenicline hydrochloride is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs). This compound is intended for research use only and is not sold to patients. It demonstrates selectivity for α7 nAChRs and has shown promising results in restoring memory function in preclinical studies.
  • Novel partial agonist of α7 neuronal nicotinic acetylcholine receptors
  • Purity of 98.01%
  • White to off-white solid appearance
  • Exhibits good brain penetration
  • Restores memory function in scopolamine-treated rats
  • Moderately binds to rat plasma proteins