Learn More
Medchemexpress LLC Encenicline hydrochloride | 550999-74-1 | C16H18Cl2N2OS | 25 MG
SDP
Catalog No. 5000375214
Encompass_Preferred
Encenicline hydrochloride is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs), primarily for research use. It exhibits high selectivity for α7 nAChRs and demonstrates good brain penetration.
  • Acts as a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors
  • Exhibits selectivity for α7 nAChRs
  • Demonstrates good brain penetration and adequate exposure time
  • Restores memory function in scopolamine-treated rats
  • Investigated in clinical trials for conditions like Alzheimer's disease, dementia, and schizophrenia
Catalog No. 50-003-75214 Supplier Medchemexpress LLC Supplier No. 25MGHY15430A
Encenicline hydrochloride is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs), primarily for research use. It exhibits high selectivity for α7 nAChRs and demonstrates good brain penetration.
  • Acts as a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors
  • Exhibits selectivity for α7 nAChRs
  • Demonstrates good brain penetration and adequate exposure time
  • Restores memory function in scopolamine-treated rats
  • Investigated in clinical trials for conditions like Alzheimer's disease, dementia, and schizophrenia