Learn More
ATCC Blastocystis hominis; BT1
Catalog No. 502380783
Encompass
Blastocystis hominis strain BT1 is a parasitic protozoan that was isolated in 1986 from human feces. This product has applications in enteric disease research.
Catalog No. 50-238-0783 Supplier ATCC Supplier No. 50608
Blastocystis hominis strain BT1 is a parasitic protozoan that was isolated in 1986 from human feces. This product has applications in enteric disease research.