Learn More
ATCC Encephalitozoon intestinalis; alveolar isolate
Catalog No. 502379854
Encompass
Encephalitozoon intestinalis was isolated from the alveolar material of an HIV-seropositive patient. This parasitic protozoan can be cultivated in WI-38 (ATCC CCL-75), MDCK (NBL-2) (ATCC CCL-34), or BS-C-1 (ATCC CCL-26) cells and can be used in enteric disease research.
Catalog No. 50-237-9854 Supplier ATCC Supplier No. 50506
Encephalitozoon intestinalis was isolated from the alveolar material of an HIV-seropositive patient. This parasitic protozoan can be cultivated in WI-38 (ATCC CCL-75), MDCK (NBL-2) (ATCC CCL-34), or BS-C-1 (ATCC CCL-26) cells and can be used in enteric disease research.