Learn More
ATCC Encephalomyocarditis; virus
Catalog No. 502384436
Encompass
Encephalomyocarditis virus is cell culture-adapted and can be propagated in Vero cells (ATCC CCL-81).
Catalog No. 50-238-4436 Supplier ATCC Supplier No. VR1762
Encephalomyocarditis virus is cell culture-adapted and can be propagated in Vero cells (ATCC CCL-81).