Learn More
ATCC Encephalomyocarditis virus; Strain: EMC
Catalog No. 502383313
Encompass
Encephalomyocarditis virus strain EMC is propagated in Vero cells (ATCC CCL-81). The strain was isolated from a chimpanzee in Florida in 1944 and is indistinguishable antigenically from strain MM and Mengo encephalomyocarditis virus.
Catalog No. 50-238-3313 Supplier ATCC Supplier No. VR129B
Encephalomyocarditis virus strain EMC is propagated in Vero cells (ATCC CCL-81). The strain was isolated from a chimpanzee in Florida in 1944 and is indistinguishable antigenically from strain MM and Mengo encephalomyocarditis virus.