Learn More
TARGETMOL CHEMICALS INC Encequidar mesylate 25MG
SDP
Catalog No. 502880786
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Encequidar mesylate (HM30181A mesylate) is a competitive P-glycoprotein inhibitor. Purity 99.67%
Catalog No. 50-288-0786 Supplier TARGETMOL CHEMICALS INC Supplier No. T11578L25MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Encequidar mesylate (HM30181A mesylate) is a competitive P-glycoprotein inhibitor. Purity 99.67%